Search
Search

AGO1| Gene Symbol: AGO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001317212.1 | NP_001304141.1 | 16 - 17 | 2242 | 80 | |
| XM_015297696.2 | XP_015153182.1 | 15 - 16 | 2125 | 80 | ||
Set Membership: |
|