Search
Search

MAPK10| Gene Symbol: MAPK10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318224.3 | NP_001305153.1 | 3 - 4 | 537 | 117 | |
| NM_001318225.3 | NP_001305154.1 | 3 - 4 | 537 | 117 | ||
| XM_025149638.1 | XP_025005406.1 | 3 - 4 | 690 | 117 | ||
| XM_025149639.1 | XP_025005407.1 | 3 - 4 | 422 | 117 | ||
Set Membership: |
|