Search
Search

PI16| Gene Symbol: PI16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015299137.2 | XP_015154623.2 | 605 | 69 | ||
| XM_025143864.1 | XP_024999632.1 | 604 | 69 | |||
| XM_025143865.1 | XP_024999633.1 | 3286 | 69 | |||
| XM_025143866.1 | XP_024999634.1 | 3561 | 69 | |||
| XM_025143867.1 | XP_024999635.1 | 2977 | 69 | |||
| GenBank mRNA | CR338830.1 | - | 563 | 69 | ||
Set Membership: |
|