Search
Search

RBPJL| Gene Symbol: RBPJL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004947026.3 | XP_004947083.1 | 8 - 9 | 1053 | 100 | |
| XM_015296737.2 | XP_015152223.1 | 8 - 9 | 1057 | 100 | ||
| XM_025142338.1 | XP_024998106.1 | 9 - 10 | 4979 | 100 | ||
| XM_417363.6 | XP_417363.3 | 8 - 9 | 1048 | 100 | ||
| GenBank mRNA | CR338982.2 | - | 8 - 9 | 959 | 100 | |
Set Membership: |
|