Search
Search

SNRNP25| Gene Symbol: SNRNP25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015294611.1 | XP_015150097.1 | 5 - 6 | 750 | 112 | |
| GenBank mRNA | BX929371.2 | - | 3 - 4 | 334 | 112 | |
| BX934925.1 | - | 5 - 6 | 599 | 112 | ||
| CR338802.2 | - | 6 - 7 | 830 | 112 | ||
| CR406045.1 | - | 4 - 5 | 540 | 112 | ||
Set Membership: |
|