Search
Search

CDK7| Gene Symbol: CDK7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001234586.5 | XP_001234587.2 | 9 - 10 | 746 | 78 | |
| GenBank mRNA | BX929547.2 | - | 9 - 10 | 822 | 78 | |
| BX930305.1 | - | 8 - 9 | 609 | 78 | ||
Set Membership: |
|