Search
Search

NDUFS2| Gene Symbol: NDUFS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015280055.1 | XP_015135541.1 | 8 - 9 | 958 | 51 | |
| GenBank mRNA | BX929653.1 | - | 6 - 7 | 618 | 51 | |
| BX934559.1 | - | 8 - 9 | 865 | 51 | ||
| CR524210.1 | - | 6 - 7 | 654 | 51 | ||
Set Membership: |
|