Search
Search

RUBCN| Gene Symbol: RUBCN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015291463.2 | XP_015146949.1 | 23 | 4265 | 153 | |
| XM_015291464.2 | XP_015146950.1 | 22 | 4220 | 153 | ||
| XM_015291465.2 | XP_015146951.1 | 22 | 4190 | 153 | ||
| XM_015291466.2 | XP_015146952.1 | 21 | 4144 | 153 | ||
| XM_015291467.2 | XP_015146953.1 | 23 | 4325 | 153 | ||
| GenBank mRNA | BX929783.2 | - | 1 | 214 | 155 | |
Set Membership: |
|