Search
Search

WDR74| Gene Symbol: WDR74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015273030.2 | XP_015128516.2 | 9 - 10 | 987 | 63 | |
| GenBank mRNA | BX929806.1 | - | 9 - 10 | 916 | 63 | |
| BX933124.1 | - | 8 - 9 | 789 | 63 | ||
| CR406521.1 | - | 6 - 7 | 673 | 63 | ||
Set Membership: |
|