Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Gg07164928_s1
          See other IGFN1 GE Assays ›
          Gene Symbol:
          IGFN1
          Gene Name:
          immunoglobulin-like and fibronectin type III domain containing 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.26: 869620 - 898303 on Build GRCg6a
          Species:
          Chicken
          Species Specific ID (Flybase ID):
          -
          Assay ID Gg07164928_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: IGFN1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_015298822.2 XP_015154308.1 2607 83
          XM_015298862.2 XP_015154348.1 2607 83
          XM_015298864.2 XP_015154350.1 2607 83
          XM_025143720.1 XP_024999488.1 2607 83
          XM_025143721.1 XP_024999489.1 2631 83
          XM_025143722.1 XP_024999490.1 2590 83
          XM_025143723.1 XP_024999491.1 2607 83
          XM_025143724.1 XP_024999492.1 2607 83
          XM_025143725.1 XP_024999493.1 2607 83
          XM_025143726.1 XP_024999494.1 2607 83
          XM_025143727.1 XP_024999495.1 2607 83
          XM_025143728.1 XP_024999496.1 2607 83
          XM_025143729.1 XP_024999497.1 2607 83
          XM_025143730.1 XP_024999498.1 2607 83
          XM_025143731.1 XP_024999499.1 2607 83
          XM_025143732.1 XP_024999500.1 2607 83
          XM_025143733.1 XP_024999501.1 2607 83
          XM_025143734.1 XP_024999502.1 2607 83
          XM_025143735.1 XP_024999503.1 2607 83
          XM_025143736.1 XP_024999504.1 2607 83
          XM_025143737.1 XP_024999505.1 2607 83
          XM_025143738.1 XP_024999506.1 2607 83
          XM_025143739.1 XP_024999507.1 2607 83
          XM_025143741.1 XP_024999509.1 2607 83
          XM_025143742.1 XP_024999510.1 2607 83
          XM_025143743.1 XP_024999511.1 2607 83
          XM_025143744.1 XP_024999512.1 2607 83
          XM_025143745.1 XP_024999513.1 2607 83
          XM_025143746.1 XP_024999514.1 2607 83
          XM_025143747.1 XP_024999515.1 2607 83
          XM_025143748.1 XP_024999516.1 2607 83
          XM_025143749.1 XP_024999517.1 2607 83
          XM_025143750.1 XP_024999518.1 2607 83
          XM_025143751.1 XP_024999519.1 2607 83
          XM_025143752.1 XP_024999520.1 2607 83
          XM_025143753.1 XP_024999521.1 2607 83
          XM_025143754.1 XP_024999522.1 2607 83
          XM_025143755.1 XP_024999523.1 2607 83
          XM_025143757.1 XP_024999525.1 2607 83
          XM_025143758.1 XP_024999526.1 2607 83
          XM_025143759.1 XP_024999527.1 2607 83
          XM_025143760.1 XP_024999528.1 2607 83
          XM_025143761.1 XP_024999529.1 2607 83
          XM_025143762.1 XP_024999530.1 2607 83
          XM_025143763.1 XP_024999531.1 2607 83
          XM_025143764.1 XP_024999532.1 2607 83
          XM_025143765.1 XP_024999533.1 2607 83
          XM_025143766.1 XP_024999534.1 2607 83
          XM_025143767.1 XP_024999535.1 2607 83
          XM_025143768.1 XP_024999536.1 2607 83
          GenBank mRNA CR354181.1 - 523 83

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline