Search
Search

SMG6| Gene Symbol: SMG6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004946547.3 | XP_004946604.2 | 19 | 100 | ||
| XM_004946548.3 | XP_004946605.2 | 19 | 100 | |||
| GenBank mRNA | CR385697.1 | - | 1 | 621 | 101 | |
Set Membership: |
|