Search
Search

SSH2| Gene Symbol: SSH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015296101.2 | XP_015151587.2 | 15 | 78 | ||
| XM_015296103.2 | XP_015151589.1 | 13 | 78 | |||
| XM_415832.6 | XP_415832.5 | 14 | 78 | |||
| GenBank mRNA | CR386096.1 | - | 1 | 626 | 78 | |
Set Membership: |
|