Search
Search

GOLGA4| Gene Symbol: GOLGA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015281656.2 | XP_015137142.1 | 25 - 26 | 8725 | 112 | |
| XM_015281658.2 | XP_015137144.1 | 24 - 25 | 8626 | 112 | ||
| XM_015281659.2 | XP_015137145.1 | 25 - 26 | 6935 | 112 | ||
| XM_015281660.2 | XP_015137146.1 | 24 - 25 | 6791 | 112 | ||
| GenBank mRNA | CR386232.1 | - | 1 - 2 | 372 | 112 | |
Set Membership: |
|