Search
Search

SLC37A1| Gene Symbol: SLC37A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015300960.2 | XP_015156446.1 | 21 | 4690 | 98 | |
| XM_015300966.2 | XP_015156452.1 | 21 | 4661 | 98 | ||
| XM_015300972.2 | XP_015156458.1 | 22 | 4809 | 98 | ||
| XM_015300978.2 | XP_015156464.1 | 21 | 7546 | 98 | ||
| XM_025145568.1 | XP_025001336.1 | 21 | 4694 | 98 | ||
| GenBank mRNA | CR386601.1 | - | 1 | 1123 | 98 | |
Set Membership: |
|