Search
Search

GRK3| Gene Symbol: GRK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015275493.2 | XP_015130979.1 | 9 - 10 | 828 | 108 | |
| XM_015275494.2 | XP_015130980.1 | 9 - 10 | 768 | 108 | ||
| GenBank mRNA | CR405860.1 | - | 9 - 10 | 692 | 108 | |
Set Membership: |
|