Search
Search

PIGG| Gene Symbol: PIGG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004949326.3 | XP_004949383.1 | 6 - 7 | 1188 | 145 | |
| XM_004949327.3 | XP_004949384.1 | 6 - 7 | 1188 | 145 | ||
| XM_004949328.3 | XP_004949385.1 | 6 - 7 | 1188 | 145 | ||
| XM_015280488.2 | XP_015135974.1 | 6 - 7 | 1187 | 145 | ||
| XM_025144839.1 | XP_025000607.1 | 2 - 3 | 309 | 145 | ||
| XM_424872.6 | XP_424872.3 | 6 - 7 | 1187 | 145 | ||
| GenBank mRNA | CR406905.1 | - | 6 - 7 | 996 | 145 | |
Set Membership: |
|