Search
Search

CFAP70| Gene Symbol: CFAP70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001232753.5 | XP_001232754.3 | 21 - 22 | 2954 | 113 | |
| XM_015294694.2 | XP_015150180.1 | 21 - 22 | 2954 | 113 | ||
| XM_015294695.2 | XP_015150181.1 | 20 - 21 | 2873 | 113 | ||
| XM_015294696.2 | XP_015150182.1 | 21 - 22 | 2954 | 113 | ||
| XM_015294697.2 | XP_015150183.1 | 20 - 21 | 2777 | 113 | ||
| XM_015294698.1 | XP_015150184.1 | 21 - 22 | 2613 | 113 | ||
| XM_015294700.2 | XP_015150186.1 | 20 - 21 | 2468 | 113 | ||
| XM_025155392.1 | XP_025011160.1 | 21 - 22 | 2954 | 113 | ||
| GenBank mRNA | CR407181.1 | - | 1 - 2 | 68 | 113 | |
Set Membership: |
|