Search
Search

UBTF| Gene Symbol: UBTF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015273207.2 | XP_015128693.1 | 5 - 6 | 536 | 72 | |
| XM_015273208.1 | XP_015128694.1 | 5 - 6 | 536 | 72 | ||
| XM_025144211.1 | XP_024999979.1 | 6 - 7 | 864 | 72 | ||
| XM_025144212.1 | XP_024999980.1 | 6 - 7 | 980 | 72 | ||
| XM_025144213.1 | XP_024999981.1 | 6 - 7 | 626 | 72 | ||
| XM_025144214.1 | XP_024999982.1 | 6 - 7 | 626 | 72 | ||
| XM_025144215.1 | XP_024999983.1 | 6 - 7 | 625 | 72 | ||
| XM_025144216.1 | XP_024999984.1 | 6 - 7 | 625 | 72 | ||
| XM_025144217.1 | XP_024999985.1 | 6 - 7 | 863 | 72 | ||
| GenBank mRNA | CR522893.1 | - | 4 - 5 | 413 | 72 | |
Set Membership: |
|