Search
Search

TMEM196| Gene Symbol: TMEM196 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001234721.5 | XP_001234722.2 | 3 - 4 | 585 | 103 | |
| XM_004939341.3 | XP_004939398.1 | 3 - 4 | 643 | 103 | ||
| XM_025147412.1 | XP_025003180.1 | 3 - 4 | 483 | 103 | ||
| XM_025147413.1 | XP_025003181.1 | 2 - 3 | 491 | 103 | ||
| GenBank mRNA | CR523746.1 | - | 2 - 3 | 313 | 103 | |
Set Membership: |
|