Search
Search

CCDC83| Gene Symbol: CCDC83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015280774.2 | XP_015136260.1 | 8 - 9 | 937 | 88 | |
| XM_015280783.2 | XP_015136269.1 | 8 - 9 | 937 | 88 | ||
| XM_025146923.1 | XP_025002691.1 | 9 - 10 | 1141 | 88 | ||
| XM_423016.6 | XP_423016.4 | 8 - 9 | 937 | 88 | ||
| GenBank mRNA | CR733273.1 | - | 8 - 9 | 938 | 88 | |
Set Membership: |
|