Search
Search

DDC| Gene Symbol: DDC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004935144.3 | XP_004935201.1 | 14 | 2940 | 64 | |
| XM_015282289.2 | XP_015137775.1 | 14 | 3738 | 64 | ||
| XM_419032.6 | XP_419032.3 | 14 | 2979 | 64 | ||
Set Membership: |
|