Search
Search

CDC23| Gene Symbol: CDC23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003642103.4 | XP_003642151.1 | 10 - 11 | 1153 | 102 | |
| XM_025155034.1 | XP_025010802.1 | 8 - 9 | 1157 | 102 | ||
Set Membership: |
|