Search
Search

BTRC| Gene Symbol: BTRC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015288763.2 | XP_015144249.1 | 3 - 4 | 322 | 141 | |
| XM_015288765.2 | XP_015144251.1 | 2 - 3 | 253 | 141 | ||
| XM_015288767.2 | XP_015144253.1 | 3 - 4 | 322 | 141 | ||
Set Membership: |
|