Search
Search

RAB3GAP1| Gene Symbol: RAB3GAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003641627.4 | XP_003641675.2 | 21 - 22 | 2707 | 69 | |
| XM_422135.6 | XP_422135.4 | 21 - 22 | 2506 | 69 | ||
Set Membership: |
|