Search
Search

UBE2Q1| Gene Symbol: UBE2Q1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025143464.1 | XP_024999232.1 | 3 - 4 | 88 | ||
| GenBank mRNA | BX930382.1 | - | 3 - 4 | 307 | 88 | |
| BX933193.1 | - | 2 - 3 | 149 | 88 | ||
| BX934095.1 | - | 3 - 4 | 388 | 88 | ||
Set Membership: |
|