Search
Search

IFT43| Gene Symbol: IFT43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004941812.3 | XP_004941869.1 | 4 - 5 | 288 | 118 | |
| XM_015287703.2 | XP_015143189.1 | 5 - 6 | 330 | 118 | ||
| XM_015287704.2 | XP_015143190.1 | 4 - 5 | 299 | 118 | ||
| XM_015287705.1 | XP_015143191.1 | 5 - 6 | 444 | 118 | ||
| GenBank mRNA | BX930803.2 | - | 4 - 5 | 237 | 118 | |
| BX930829.1 | - | 4 - 5 | 239 | 118 | ||
| BX931067.2 | - | 4 - 5 | 241 | 118 | ||
| BX932188.1 | - | 3 - 4 | 341 | 118 | ||
Set Membership: |
|