Search
Search

MTX2| Gene Symbol: MTX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015289585.2 | XP_015145071.1 | 7 - 8 | 573 | 121 | |
| XM_421989.5 | XP_421989.1 | 9 - 10 | 689 | 121 | ||
| GenBank mRNA | BX930870.2 | - | 6 - 7 | 423 | 121 | |
| BX933987.2 | - | 9 - 10 | 658 | 121 | ||
Set Membership: |
|