Search
Search

PPEF2| Gene Symbol: PPEF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015276420.1 | XP_015131906.1 | 11 - 12 | 3855 | 92 | |
| XM_015276422.1 | XP_015131908.1 | 11 - 12 | 8592 | 92 | ||
Set Membership: |
|