Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ?
立即註冊
產品
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Greener Products
查看所有產品類別
應用
生物製程
細胞培養和轉染
食品和飲料
實驗室解決方案
製藥和生物製藥
即時定量PCR
半導體分析
臨床和診斷
數位解決方案
分子光譜、同位素與元素分析
查看所有應用與技術
服務
360° CDMO 和 CRO 服務
CDMO 服务
临床试验 CRO 服务
客製化服務
金融與租賃服務
儀器服務
實驗室資訊學
授權與商業供應
培訓服務
查看所有服務
支援
線上訂購協助
數位解決方案
產品支持
技術相關資訊
培訓與教育
查看所有協助與支援
熱門商品
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
我們的服務對象
生物技術
生物製藥
CDMO
實驗室診斷
工業與應用科學
特價商品
聯絡我們
快速訂購
訂單狀態與貨物追蹤
文件和證書
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
訂單查詢
快速訂購
登入
登入
還沒有帳戶嗎 ?
立即註冊
帳戶
訂單
Connect: Lab, Data, Apps
客製化產品與項目
儀器管理
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Gg07175779_m1
See other RSAD1 GE Assays ›
Gene Symbol:
RSAD1
Gene Name:
radical S-adenosyl methionine domain containing 1
Gene Aliases:
MYCBPAP
Chromosome Location:
Chr.18: 10323045 - 10337930 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID
Gg07175779_m1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
新增到購物車
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: RSAD1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_015295489.2
XP_015150975.1
10 - 11
1649
93
XM_015295490.2
XP_015150976.1
10 - 11
1646
93
XM_015295491.2
XP_015150977.1
11 - 12
1558
93
XM_015295492.2
XP_015150978.1
10 - 11
1647
93
XM_015295493.2
XP_015150979.1
10 - 11
1648
93
XM_015295494.2
XP_015150980.1
10 - 11
1648
93
XM_015295495.2
XP_015150981.1
10 - 11
1648
93
XM_015295496.2
XP_015150982.1
10 - 11
1649
93
XM_015295497.2
XP_015150983.1
10 - 11
1649
93
XM_015295498.2
XP_015150984.1
7 - 8
1155
93
XM_015295500.2
XP_015150986.1
10 - 11
1653
93
XM_015295502.2
XP_015150988.1
7 - 8
1084
93
XM_015295503.2
XP_015150989.1
7 - 8
1075
93
XM_015295504.2
XP_015150990.1
3 - 4
1492
93
XM_025141841.1
XP_024997609.1
10 - 11
1650
93
XM_025141843.1
XP_024997611.1
8 - 9
1060
93
XM_025141844.1
XP_024997612.1
6 - 7
1600
93
XM_025141845.1
XP_024997613.1
7 - 8
1164
93
XM_025141846.1
XP_024997614.1
10 - 11
1655
93
XM_025141847.1
XP_024997615.1
3 - 4
1234
93
Back To Top
More Information
Set Membership:
Amplicon length between 86 and 100
3' Most
Recommended Assay
Gallus gallus
Probe spans exons
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline