Search
Search

PITPNM3| Gene Symbol: PITPNM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015296205.2 | XP_015151691.1 | 18 - 19 | 2722 | 97 | |
| XM_015296206.2 | XP_015151692.1 | 17 - 18 | 2659 | 97 | ||
| XM_015296207.2 | XP_015151693.1 | 18 - 19 | 2583 | 97 | ||
| XM_015296208.2 | XP_015151694.1 | 18 - 19 | 2856 | 97 | ||
| XM_415929.6 | XP_415929.3 | 18 - 19 | 2776 | 97 | ||
Set Membership: |
|