Search
Search

NUDT12| Gene Symbol: NUDT12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001231286.5 | XP_001231287.1 | 7 | 1862 | 100 | |
| XM_004949297.3 | XP_004949354.1 | 7 | 1943 | 100 | ||
| XM_015280457.2 | XP_015135943.1 | 7 | 1905 | 100 | ||
| XM_025145017.1 | XP_025000785.1 | 7 | 1939 | 100 | ||
| XM_025145018.1 | XP_025000786.1 | 7 | 1906 | 100 | ||
Set Membership: |
|