Search
Search

CACNA1I| Gene Symbol: CACNA1I | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015288724.2 | XP_015144210.2 | 25 - 26 | 5531 | 119 | |
| XM_015288734.2 | XP_015144220.2 | 25 - 26 | 5531 | 119 | ||
Set Membership: |
|