Search
Search

RSPRY1| Gene Symbol: RSPRY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004944057.3 | XP_004944114.1 | 16 - 17 | 2118 | 87 | |
| XM_004944058.3 | XP_004944115.1 | 15 - 16 | 2093 | 87 | ||
| XM_015292336.2 | XP_015147822.1 | 16 - 17 | 2099 | 87 | ||
| XM_015292337.2 | XP_015147823.1 | 15 - 16 | 1622 | 87 | ||
| XM_025154260.1 | XP_025010028.1 | 15 - 16 | 2610 | 87 | ||
Set Membership: |
|