Search
Search

BRAT1| Gene Symbol: BRAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001233205.5 | XP_001233206.3 | 1 - 2 | 367 | 119 | |
| XM_004945143.3 | XP_004945200.1 | 1 - 2 | 367 | 119 | ||
Set Membership: |
|