Search
Search

SLC24A3| Gene Symbol: SLC24A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004940143.3 | XP_004940200.1 | 13 - 14 | 5681 | 98 | |
| XM_015283392.2 | XP_015138878.1 | 13 - 14 | 1528 | 98 | ||
| XM_015283393.2 | XP_015138879.1 | 13 - 14 | 1516 | 98 | ||
Set Membership: |
|