Search
Search

CARD10| Gene Symbol: CARD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015288372.2 | XP_015143858.1 | 20 - 21 | 5669 | 103 | |
| XM_015288374.2 | XP_015143860.1 | 20 - 21 | 5630 | 103 | ||
| XM_015288378.2 | XP_015143864.1 | 20 - 21 | 5589 | 103 | ||
| XM_015288385.2 | XP_015143871.1 | 18 - 19 | 2829 | 103 | ||
Set Membership: |
|