Search
Search

CARD10| Gene Symbol: CARD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015288372.2 | XP_015143858.1 | 21 - 22 | 5786 | 162 | |
| XM_015288374.2 | XP_015143860.1 | 21 - 22 | 5747 | 162 | ||
| XM_015288378.2 | XP_015143864.1 | 21 - 22 | 5706 | 162 | ||
| XM_015288385.2 | XP_015143871.1 | 19 - 20 | 2946 | 162 | ||
Set Membership: |
|