Search
Search

CAPN13| Gene Symbol: CAPN13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004940180.3 | XP_004940237.2 | 19 - 20 | 2093 | 58 | |
| XM_025149358.1 | XP_025005126.1 | 18 - 19 | 2075 | 58 | ||
Set Membership: |
|