Search
Search

CAPN10| Gene Symbol: CAPN10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015291569.2 | XP_015147055.1 | 7 - 8 | 1024 | 77 | |
| XM_025153505.1 | XP_025009273.1 | 8 - 9 | 5486 | 77 | ||
Set Membership: |
|