Search
Search

LOC107055361| Gene Symbol: LOC107055361 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015300113.2 | XP_015155599.1 | 2 - 3 | 553 | 66 | |
| GenBank mRNA | MN326300.1 | - | 2 - 3 | 478 | 66 | |
Set Membership: |
|