Search
Search

CTPS1| Gene Symbol: CTPS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004947692.3 | XP_004947749.3 | 17 - 18 | 1994 | 119 | |
| XM_004947693.3 | XP_004947750.1 | 17 - 18 | 2007 | 119 | ||
| XM_025143037.1 | XP_024998805.1 | 18 - 19 | 1884 | 119 | ||
Set Membership: |
|