Search
Search

DIS3L2| Gene Symbol: DIS3L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015291462.2 | XP_015146948.2 | 18 - 19 | 3111 | 134 | |
| XM_025153426.1 | XP_025009194.1 | 18 - 19 | 2568 | 134 | ||
| XM_025153427.1 | XP_025009195.1 | 18 - 19 | 2526 | 134 | ||
Set Membership: |
|