Search
Search

CEP131| Gene Symbol: CEP131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015295620.1 | XP_015151106.1 | 14 - 15 | 2001 | 70 | |
| XM_025141707.1 | XP_024997475.1 | 14 - 15 | 2001 | 70 | ||
| XM_025141708.1 | XP_024997476.1 | 14 - 15 | 1980 | 70 | ||
| XM_025141709.1 | XP_024997477.1 | 14 - 15 | 2001 | 70 | ||
Set Membership: |
|