Search
Search

SRR| Gene Symbol: SRR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001234890.5 | XP_001234891.1 | 5 - 6 | 628 | 70 | |
| XM_004946542.3 | XP_004946599.1 | 5 - 6 | 633 | 70 | ||
| XM_015296054.2 | XP_015151540.1 | 4 - 5 | 691 | 70 | ||
| XM_015296055.2 | XP_015151541.1 | 4 - 5 | 698 | 70 | ||
| XM_015296056.2 | XP_015151542.1 | 4 - 5 | 704 | 70 | ||
| XM_025142068.1 | XP_024997836.1 | 4 - 5 | 618 | 70 | ||
Set Membership: |
|