Search
Search

PHF19| Gene Symbol: PHF19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015279816.2 | XP_015135302.1 | 15 | 3557 | 98 | |
| XM_015279817.2 | XP_015135303.1 | 15 | 3504 | 98 | ||
| XM_015279818.2 | XP_015135304.1 | 15 | 3374 | 98 | ||
| XM_025141497.1 | XP_024997265.1 | 15 | 3677 | 98 | ||
Set Membership: |
|