Search
Search

HDGF| Gene Symbol: HDGF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001236426.5 | XP_001236427.2 | 2 - 3 | 265 | 122 | |
| GenBank mRNA | BX932287.1 | - | 2 - 3 | 222 | 122 | |
| BX932332.2 | - | 2 - 3 | 218 | 122 | ||
| BX935897.1 | - | 2 - 3 | 128 | 122 | ||
Set Membership: |
|