Search
Search

RBFOX2| Gene Symbol: RBFOX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015288005.2 | XP_015143491.1 | 11 - 12 | 1399 | 68 | |
| XM_015288009.2 | XP_015143495.1 | 11 - 12 | 1401 | 68 | ||
| XM_025155472.1 | XP_025011240.1 | 10 - 11 | 1149 | 68 | ||
| XM_025155479.1 | XP_025011247.1 | 10 - 11 | 1154 | 68 | ||
| XM_025155504.1 | XP_025011272.1 | 9 - 10 | 1058 | 68 | ||
| GenBank mRNA | BX932391.2 | - | 11 - 12 | 1048 | 68 | |
Set Membership: |
|