Search
Search

FOXI2| Gene Symbol: FOXI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003641507.4 | XP_003641555.4 | 1 - 2 | 562 | 102 | |
| XM_015289046.2 | XP_015144532.2 | 1 - 2 | 564 | 102 | ||
| GenBank mRNA | BX933133.1 | - | 6 - 7 | 736 | 102 | |
Set Membership: |
|