Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Gg07185338_g1
          See other INPP4A GE Assays ›
          Gene Symbol:
          INPP4A
          Gene Name:
          inositol polyphosphate-4-phosphatase type I A
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 133488998 - 133602459 on Build GRCg6a
          Species:
          Chicken
          Species Specific ID (Flybase ID):
          -
          Assay ID Gg07185338_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: INPP4A
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_004938477.3 XP_004938534.1 14 - 15 1649 68
          XM_004938478.3 XP_004938535.1 14 - 15 1637 68
          XM_004938480.3 XP_004938537.1 14 - 15 1649 68
          XM_004938482.3 XP_004938539.1 14 - 15 1606 68
          XM_004938484.3 XP_004938541.1 14 - 15 1645 68
          XM_004938485.3 XP_004938542.1 14 - 15 1603 68
          XM_004938486.2 XP_004938543.2 11 - 12 1222 68
          XM_004938487.3 XP_004938544.1 14 - 15 1603 68
          XM_004938488.3 XP_004938545.1 14 - 15 1647 68
          XM_004938489.3 XP_004938546.1 14 - 15 1648 68
          XM_004938490.3 XP_004938547.1 14 - 15 1605 68
          XM_004938491.3 XP_004938548.1 14 - 15 1601 68
          XM_004938492.3 XP_004938549.1 14 - 15 1601 68
          XM_015277773.2 XP_015133259.1 14 - 15 1551 68
          XM_015277774.1 XP_015133260.1 13 - 14 1556 68
          XM_015277775.2 XP_015133261.1 13 - 14 1435 68
          XM_015277776.1 XP_015133262.1 12 - 13 1416 68
          XM_015277777.2 XP_015133263.1 13 - 14 1509 68
          XM_015277778.2 XP_015133264.1 12 - 13 1294 68
          XM_015277780.2 XP_015133266.1 14 - 15 1649 68
          XM_015277781.1 XP_015133267.1 12 - 13 1374 68
          XM_025146639.1 XP_025002407.1 14 - 15 1656 68
          XM_025146640.1 XP_025002408.1 14 - 15 1606 68
          XM_025146641.1 XP_025002409.1 14 - 15 1604 68
          XM_025146642.1 XP_025002410.1 14 - 15 1603 68
          XM_416886.5 XP_416886.2 14 - 15 1602 68
          GenBank mRNA BX934602.2 - 11 - 12 1154 68

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Gallus gallus Probe spans exons

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline