Search
Search

CCDC27| Gene Symbol: CCDC27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004947364.3 | XP_004947421.1 | 2 - 3 | 1362 | 88 | |
| XM_004947366.3 | XP_004947423.1 | 2 - 3 | 1326 | 88 | ||
| XM_015297094.2 | XP_015152580.1 | 2 - 3 | 1365 | 88 | ||
| XM_025142633.1 | XP_024998401.1 | 2 - 3 | 1329 | 88 | ||
| GenBank mRNA | BX935428.2 | - | 1 - 2 | 90 | 88 | |
Set Membership: |
|